Please login to your online account to display your discounted pricing

gamma delta TCR Armenian Hamster anti-Mouse, PerCP-eFluor 710, Clone: GL-3, eBioscience™

Armenian Hamster Monoclonal Antibody

$162.40 - $456.75

Specifications

Antigen gamma delta TCR
Clone GL3)
Host Species Armenian Hamster
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016067

 
life technologies
LS46571180
25μg Each for $162.40

5016068

 
life technologies
LS46571182
100μg Each for $456.75
Description & Specifications

Specifications

Antigen gamma delta TCR
Clone GL3)
Host Species Armenian Hamster
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The GL3 monoclonal antibody reacts with the gamma delta T-cell Receptor complex (TCR) on all gamma delta TCR bearing T cells, but it does not react with alpha beta TCR. The gamma delta TCR is present on cells in the thymus, epidermis, epithelial lining of the intestine, peritoneal cavity, and lymphoid tissues.