missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

gamma delta TCR Armenian Hamster anti-Mouse, PE-Cy7, Clone: GL-3, eBioscience™

Armenian Hamster Monoclonal Antibody

$159.50 - $397.30

Specifications

Antigen gamma delta TCR
Clone GL3)
Host Species Armenian Hamster
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123382 Life Technologies
LS25571180
25μg Each for $159.50
501123383 Life Technologies
LS25571182
100μg Each for $397.30
Specifications

Description

The GL3 monoclonal antibody reacts with the gamma delta T-cell Receptor complex (TCR) on all gamma delta TCR bearing T cells, but it does not react with alpha beta TCR. The gamma delta TCR is present on cells in the thymus, epidermis, epithelial lining of the intestine, peritoneal cavity, and lymphoid tissues.
Description & Specifications

Specifications

gamma delta TCR
Armenian Hamster
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL
GL3)
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer