missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

gamma delta TCR Armenian Hamster anti-Mouse, FITC, Clone: UC7-13D5, eBioscience™

Armenian Hamster Monoclonal Antibody

$104.28 - $381.48

Specifications

Antigen gamma delta TCR
Clone UC7-13D5
Host Species Armenian Hamster
Gene Alias gd TCR, gdTCR, TCRgd
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122142 Life Technologies
LS11581182
100μg Each for $236.28
509866 Life Technologies
LS11581181
50μg Each for $104.28
509868 Life Technologies
LS11581185
500μg Each for $381.48
Specifications

Description

The UC7-13D5 monoclonal antibody reacts with the mouse γδ TCR complex. γδ TCR is expressed by a small subset of T cells in the thymus, peripheral lymphoid tissues, intestinal epithelium, and epidermis. The exact specificity, ligand and function of γδ TCR-bearing T cells are not yet fully understood; it is suggested that these cells recognize bacterial ligands and some tumor cells in the context of MHC class I-like gene products and play a role in regulation of the immune response and during bacterial infection.
Description & Specifications

Specifications

gamma delta TCR
Armenian Hamster
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
UC7-13D5
gd TCR, gdTCR, TCRgd
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL