Please login to your online account to display your discounted pricing

Alfa Aesar™ gamma-Bag Cell Peptide


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66911MA

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66911MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Storage Requirements -30° to -10°C
Molecular Weight 705.8
Purification Method HPLC
Regulatory Status RUO
Sequence Arg-Leu-Arg-Phe-Asp
Product Type gamma-Bag Cell Peptide
Quantity 10mg
Format Solid