Please login to your online account to display your discounted pricing

Alfa Aesar™ gamma-Bag Cell Peptide

For cellular and molecular biology applications

Specifications

Product Type gamma-Bag Cell Peptide
Molecular Weight 705.8
Storage Requirements -30° to -10°C
Format Solid
Sequence Arg-Leu-Arg-Phe-Asp
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66911MA

 
alfa aesar™
J66911MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66911MB

 
alfa aesar™
J66911MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type gamma-Bag Cell Peptide
Molecular Weight 705.8
Storage Requirements -30° to -10°C
Format Solid
Sequence Arg-Leu-Arg-Phe-Asp
Regulatory Status RUO
Purification Method HPLC