missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ Gambogic acid

Apoptosis inducer. Activates caspases and inhibits Bcl-2 family proteins

$145.69 - $614.42

Specifications

Product Type Gambogic acid
Inhibitors Gambogic acid
Molecular Weight 628.75
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
359010R Tocris™
RDS359010
10mg Each for $145.69
359050R Tocris™
RDS359050
50mg Each for $614.42
Specifications

Description

Natural product isolated from the Garcinia hanburyi tree. Induces apoptosis in several tumor cell lines including T47D cells. Activates caspases with an EC50 value of 0.78 - 1.64μM and competitively inhibits antiapoptotic Bcl-2 family proteins (IC50 values are 1.47, 1.21, 2.02, 0.66, 1.06 and 0.79μM for Bcl-XL, Bcl-2, Bcl-W, Bcl-B, Bfl-1 and Mcl-1 respectively). Also blocks Kir2.1 channels (EC50 ≤ 100nM).
  • Chemical Name: (2Z)-2-Methyl-4-[(1R,3aS,5S,11R,14aS)-3a,4,5,7-tetrahydro-8-hydroxy-3,3,11-trimethyl-13-(3-methyl-2-butenyl)-11-(4-methyl-3-pentenyl)-7,15-dioxo-1,5-methano-1H,3H,11H-furo[3,4-g]pyrano[3,2-b]xanthen-1-yl]-2-butenoic acid
  • Formula: C38H44O8
  • CAS Number: 2752-65-0
  • Solubility: Soluble to 75mM in DMSO and to 100mM in ethanol
Description & Specifications

Specifications

Gambogic acid
Gambogic acid
628.75