missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Galvinoxyl free radical, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 2370-18-5 |
---|---|
Molecular Formula | C29H42O2 |
Molecular Weight (g/mol) | 422.65 |
MDL Number | MFCD00001616 |
InChI Key | HRLDHROCDBYHAU-UHFFFAOYSA-N |
Synonym | 2,6-Di-tert-butyl-α-(3,5-di-tert-butyl-4-oxo-2,5-cyclohexadien-1-ylidene)-p-tolyloxy free radical |
IUPAC Name | 2,6-di-tert-butyl-4-[(3,5-di-tert-butyl-4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one |
SMILES | CC(C)(C)C1=CC(C=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC119850010
|
Thermo Fisher Scientific
119850010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC119850250
|
Thermo Fisher Scientific
AC119850250 |
25g | Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Polymerization reagentChemical Identifiers
2370-18-5 | |
422.65 | |
HRLDHROCDBYHAU-UHFFFAOYSA-N | |
2,6-di-tert-butyl-4-[(3,5-di-tert-butyl-4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one |
C29H42O2 | |
MFCD00001616 | |
2,6-Di-tert-butyl-α-(3,5-di-tert-butyl-4-oxo-2,5-cyclohexadien-1-ylidene)-p-tolyloxy free radical | |
CC(C)(C)C1=CC(C=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C |
Specifications
2370-18-5 | |
156°C to 160°C | |
Authentic | |
MFCD00001616 | |
Glass bottle | |
15, 4387 | |
1g | |
CC(C)(C)C1=CC(C=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C | |
422.65 | |
Crystalline Powder, Crystals and/or Chunks | |
Galvinoxyl free radical |
Blue to Purple | |
97.5% min. (HPLC) | |
C29H42O2 | |
08, IV, 1379 | |
01,409 | |
2,6-Di-tert-butyl-α-(3,5-di-tert-butyl-4-oxo-2,5-cyclohexadien-1-ylidene)-p-tolyloxy free radical | |
HRLDHROCDBYHAU-UHFFFAOYSA-N | |
2,6-di-tert-butyl-4-[(3,5-di-tert-butyl-4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one | |
421.64 | |
98% |
Safety and Handling
EINECSNumber : 219-133-2
RUO – Research Use Only