missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gallocyanine

$48.64 - $641.88

Chemical Identifiers

CAS 1562-85-2
Molecular Formula C15H13ClN2O5
Formula Weight 336.73
MDL Number MFCD00011926
Beilstein 307454
Merck Index 14,4355
Synonym C.I. 51030; Mordant blue 10
Chemical Name or Material Gallocyanine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1293609 SDS Alfa Aesar™
A1293609
10g Each for $48.64
AAA1293618 SDS Alfa Aesar™
A1293618
50g Each for $158.54
AAA1293630 SDS Alfa Aesar™
A1293630
250g Each for $641.88
Specifications

Description

Applications
Gallocyanine is used as an intermediate in chemical research. It is a small molecule dye that also inhibits DKK1 of the Wnt pathway.

Solubility
Soluble in water (10 mg/ml), 100% ethanol (10 mg/ml), alcohol, 1N NH4OH (10 mg/ml), and DMSO (25 mg/ml).

Notes
Keep container tightly closed in a dry and well-ventilated place. Store at room temperature. Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

1562-85-2
336.73
307454
C.I. 51030; Mordant blue 10
C15H13ClN2O5
MFCD00011926
14,4355
Gallocyanine

Specifications

10g
C15H13ClN2O5
MFCD00011926
14,4355
Gallocyanine
1562-85-2
336.73
307454
C.I. 51030; Mordant blue 10

Safety and Handling

GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P301+P310-P304+P340-P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : SP7692000