missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gallium Tin alloy, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1816103

 View more versions of this product

Catalog No. AA1816103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Gallium Tin alloy
1g
7440-31-5,7440-55-3
Sn
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
ATJFFYVFTNAWJD-UHFFFAOYSA-N
tin
5352426
Liquid
Silver
Silver-colored liquid
99.99% (metals basis)
MFCD00144390
Tin: 8.0%; Gallium: 92.0%
[Sn]
118.71
CHEBI:32990

Chemical Identifiers

7440-31-5
118.71
ATJFFYVFTNAWJD-UHFFFAOYSA-N
5352426
tin
Sn
MFCD00144390
powder, stannum, metallic, tin, elemental, wang, zinn, flake, tin, metal, silver matt powder, zinn german
CHEBI:32990
[Sn]
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

H290: May be corrosive to metals.

DOTInformation : Hazard Class: 8, Packing Group: III

SDS Documents