missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gallium, plasma standard solution, Specpure™ Ga 1000μg/mL, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 013869AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gallium | |
100 mL | |
MFCD00134045 | |
[Ga] | |
69.72 | |
CHEBI:49631 | |
23 hPa (17mm Hg) at 20°C |
7440-55-3,7647-01-0,7732-18-5 | |
Ga | |
GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
gallium | |
5360835 | |
Nitric acid: 5.0%; Gallium: 0.1%; Hydrochloric acid: <0.1%; Water: 94.8% | |
Liquid |
Chemical Identifiers
7440-55-3 | |
MFCD00134045 | |
5360835 | |
gallium |
69.72 | |
GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
CHEBI:49631 | |
[Ga] |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Hazard Class: 8, Packing Group: III
EINECSNumber : 231-714-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only