missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gallium Pellets, 6mm dia., 99.9999% (Metal Basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4432409

 View more versions of this product

Catalog No. AA4432409



Specifications

Specifications

Gallium
7440-55-3
Ga
atom
[Ga]
69.723
CHEBI:49631
10g
99.9999%, metal basis
MFCD00134045
GYHNNYVSQQEPJS-UHFFFAOYSA-N
gallium
5360835
6mm diameter Pellets

Chemical Identifiers

7440-55-3
69.723
GYHNNYVSQQEPJS-UHFFFAOYSA-N
5360835
gallium
Ga
MFCD00134045
atom
CHEBI:49631
[Ga]
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : DOT Class: 8; Packaging Group: III

EINECSNumber : 231-163-8

RTECSNumber : LW8600000

TSCA : Yes

Recommended Storage : Keep cold

Documents