missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gallium metal, 99.999% (metals basis), Alfa Aesar™

May be solid or liquid due to ambient temperature

Manufacturer:  Thermo Fisher Scientific 4441822

 View more versions of this product

Catalog No. AA4441822



Specifications

Specifications

Gallium
7440-55-3
5.91g/cm3
Ga
atom
[Ga]
69.723
CHEBI:49631
Silver to Gray
100g
Silver Gray
99.9999% (Metals basis)
MFCD00134045
GYHNNYVSQQEPJS-UHFFFAOYSA-N
gallium
5360835
May be solid or liquid due to ambient temperature

Chemical Identifiers

7440-55-3
69.723
GYHNNYVSQQEPJS-UHFFFAOYSA-N
5360835
gallium
Ga
MFCD00134045
atom
CHEBI:49631
[Ga]
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-163-8

RTECSNumber : LW8600000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents