missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gallium ingot, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1018830

 View more versions of this product

Catalog No. AA1018830



Specifications

Specifications

Gallium ingot
7440-55-3
Ga
UN2803
GYHNNYVSQQEPJS-UHFFFAOYSA-N
gallium
5360835
Ingot
Gray-Silver
250g
(metals basis)
MFCD00134045
atom, ch46oc8yv4, galio, gallium, elemental, gallium, ion ga1+, ingot, metal, metal, packaged in poylethylene bottle, pellets, 6mm dia, unii-ch46oc8yv4
[Ga]
69.723
CHEBI:49631
99.99%

Chemical Identifiers

7440-55-3
69.723
GYHNNYVSQQEPJS-UHFFFAOYSA-N
5360835
gallium
Ga
MFCD00134045
atom, ch46oc8yv4, galio, gallium, elemental, gallium, ion ga1+, ingot, metal, metal, packaged in poylethylene bottle, pellets, 6mm dia, unii-ch46oc8yv4
CHEBI:49631
[Ga]
Safety and Handling

Safety and Handling

GHS H Statement
H290
May be corrosive to metals.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: GALLIUM

EINECSNumber : 231-163-8

RTECSNumber : LW8600000

TSCA : Yes

storageNote1 : Requires overnight shipping with cold pack; Store at 2°C to 8°C

Recommended Storage : Keep cold

Documents