Gallium Indium eutectic, 99.99% (metals basis), Alfa Aesar™
Manufacturer:  Alfa Aesar 1247809
 View more versions of this product
Catalog No. AA1247809
Specifications
Specifications
Gallium Indium eutectic
99.99% (metals basis)
MFCD00144387
10g
Ga:In; 75.5:24.5 wt%
Liquid
Documents