Gallium Indium alloy, 99.99% (metals basis), Alfa Aesar™
Manufacturer:  Alfa Aesar 4424003
 View more versions of this product
Catalog No. AA4424003
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Gallium Indium alloy
1g
99.99% (metals basis)
MFCD00144387
Gray
Odorless
Ga:In; 40:60 wt.%
Solid
Documents