missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gallium Indium alloy, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4424009

 View more versions of this product

Catalog No. AA4424009

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Gallium Indium alloy
99.99% (metals basis)
MFCD00144387
Odorless
Gray
10 g
Ga:In; 40:60 wt.%
Insoluble in water.
Solid
Documents