missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gallium(III) selenide, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4557203

 View more versions of this product

Catalog No. AA4557203



Specifications

Specifications

Gallium (III) selenide
(metals basis)
MFCD00016103
376.33
99.99%
1g
12024-24-7
Ga2Se3
UN3077
Sintered Lump
3 to 12mm

Chemical Identifiers

12024-24-7
Ga2Se3
MFCD00016103
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA

SDS Documents