Please login to your online account to display your discounted pricing

Gallium(III) perchlorate hydrate, Crystalline, 13.5-15.5% Ga, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Gallium(III) perchlorate hydrate
CAS 81029-07-4
Physical Form Crystalline
Assay Percent Range 13.5-15.5% Ga
Molecular Formula Cl3GaO12·xH2O
Formula Weight 368.07g/mol (anhydrous)
MDL Number MFCD00167156
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4558004

 
alfa aesar
45580-04
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4558009

 
alfa aesar
45580-09
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 2g
Chemical Name or Material Gallium(III) perchlorate hydrate
CAS 81029-07-4
Physical Form Crystalline
Assay Percent Range 13.5-15.5% Ga
Molecular Formula Cl3GaO12·xH2O
Formula Weight 368.07g/mol (anhydrous)
MDL Number MFCD00167156
Health Hazard 1 H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
DOT Information DOT Class: 5.1, Packing Group: II
Safety and Handling

H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.