missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gallium(III) oxide, Puratronic™, 99.999% (metals basis), Alfa Aesar™

Used in vacuum deposition.

Manufacturer:  Thermo Fisher Scientific 1115106

 View more versions of this product

Catalog No. AA1115106



Specifications

Specifications

Gallium(III) oxide
Plastic bottle
6.44
5g
Ga2O3
Soluble in acids. Insoluble in water
-50 Mesh Powder
6.44
1.92
1900°C
12024-21-4
MFCD00011020
187.44
99.999%

Chemical Identifiers

12024-21-4
Ga2O3
MFCD00011020
Safety and Handling

Safety and Handling

EINECSNumber : 234-691-7

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents