missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gallium(III) oxide, 99.99% (metals basis)

In spectroscopic analysis

Manufacturer:  Alfa Aesar™ 3210214

 View more versions of this product

Catalog No. AA3210214



Specifications

Specifications

Gallium(III) oxide
99.99%
MFCD00011020
Insoluble in water; Soluble in alkalies; Very slightly soluble in hot acids
Powder
−50 Mesh
1.92
25g
12024-21-4
(metals basis)
14,4346
187.44
6.44g/mL
Odorless
1900°C

Chemical Identifiers

12024-21-4
MFCD00011020
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents