Learn More
Gallium(III) nitride, 99.99% (metals basis), Thermo Scientific Chemicals
$380.04 - $1665.27
Chemical Identifiers
CAS | 25617-97-4 |
---|---|
Molecular Formula | GaN |
Molecular Weight (g/mol) | 83.73 |
MDL Number | MFCD00016108 |
InChI Key | JMASRVWKEDWRBT-UHFFFAOYSA-N |
Synonym | gallium nitride, gallium mononitride, unii-1r9cc3p9vl, gallium nitride,, 1r9cc3p9vl, gallium nitride trace metals basis, gallium iii nitride trace metals basis 10g |
PubChem CID | 117559 |
IUPAC Name | azanylidynegallane |
SMILES | N#[Ga] |
Description
Gallium(III) nitride is used to read blu-ray discs. It is used in various wireless infrastructure applications. It finds applications in optoelectronic, high-power and high-frequency devices. It is also useful in military electronics (active electronically scanned array radars). It is biocompatible and used in electrodes and electronic components in living organisms. It is a suitable material for solar cell arrays for satellites. It is employed in bright light-emitting diodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25617-97-4 | |
83.73 | |
JMASRVWKEDWRBT-UHFFFAOYSA-N | |
117559 | |
N#[Ga] |
GaN | |
MFCD00016108 | |
gallium nitride, gallium mononitride, unii-1r9cc3p9vl, gallium nitride,, 1r9cc3p9vl, gallium nitride trace metals basis, gallium iii nitride trace metals basis 10g | |
azanylidynegallane |
Specifications
800°C (sublimation) | |
Yellow | |
10 g | |
MFCD00016108 | |
gallium nitride, gallium mononitride, unii-1r9cc3p9vl, gallium nitride,, 1r9cc3p9vl, gallium nitride trace metals basis, gallium iii nitride trace metals basis 10g | |
JMASRVWKEDWRBT-UHFFFAOYSA-N | |
azanylidynegallane | |
117559 | |
99.99% | |
6.1 g/mL | |
(metals basis) |
25617-97-4 | |
Powder | |
GaN | |
14,4351 | |
Slightly soluble in hot concentrated sulfuric acid and hot conc; sodium hydroxide; Insoluble in water and dilute acids. | |
N#[Ga] | |
83.73 | |
83.73 | |
Moisture sensitive | |
Gallium(III) nitride |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
P261-P272-P280g-P302+P352-P333+P313-P363-P501c
H317
EINECSNumber : 247-129-0
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only