missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gallium(III) nitrate solution, Ga 9-10% w/w

Manufacturer:  Alfa Aesar™ 4348418

 View more versions of this product

Catalog No. AA4348418



Specifications

Specifications

Gallium (III) nitrate solution
GaN3O9
UN3218
CHPZKNULDCNCBW-UHFFFAOYSA-N
gallium;trinitrate
61635
Liquid
50g
13494-90-1
MFCD00011016
gallium nitrate, ganite, gallium nitrate anhydrous, nitric acid, gallium salt, unii-y2v2r4w9tq, gallium iii nitrate, ccris 4493, nitric acid, gallium 3+ salt, y2v2r4w9tq, gallium salt
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ga+3]
255.735
255.73
Ga 9 to 10% w/w

Chemical Identifiers

13494-90-1
255.735
CHPZKNULDCNCBW-UHFFFAOYSA-N
61635
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ga+3]
GaN3O9
MFCD00011016
gallium nitrate, ganite, gallium nitrate anhydrous, nitric acid, gallium salt, unii-y2v2r4w9tq, gallium iii nitrate, ccris 4493, nitric acid, gallium 3+ salt, y2v2r4w9tq, gallium salt
gallium;trinitrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, AQUEOUS SOLUTION, N.O.S .

TSCA : TSCA

Documents