Learn More
Gallium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01115014
Description
Gallium(III) nitrate hydrate is the potential precursor for the synthesis of other gallium compounds. It finds application in the preparation of gallium nitride, which is used as a solar cell array for satellites. In the field of pharmaceutical industry, it is used to treat symptomatic hypercalcemia by preventing the breakdown of bone by slowing osteoclast activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gallium(III) nitrate hydrate | |
69365-72-6 | |
Crystalline Aggregates | |
(metals basis) | |
MFCD00149723 | |
14,4346 | |
Soluble in water. | |
[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
255.74 | |
255.73 (Anhydrous) | |
Puratronic™ |
110°C (decomposition) | |
White | |
25 g | |
GaN3O9 | |
UN1477 | |
gallium iii nitrate hydrate, nitric acid, gallium salt, hydrate, ga.3no3.h2o, gallium nitrate nanoparticles, gallium-iii-nitrate hydrate, gallium iii nitrate hydrate, puratronic, gallium iii nitrate hydrate, puratronic metals basis, gallium iii nitrate hydrate, crystalline trace metals basis, gallium iii nitrate hydrate, crystals and lumps trace metals basis | |
CHPZKNULDCNCBW-UHFFFAOYSA-N | |
gallium(3+) trinitrate | |
11550823 | |
99.999% | |
Hygroscopic |
Chemical Identifiers
69365-72-6 | |
255.74 | |
CHPZKNULDCNCBW-UHFFFAOYSA-N | |
11550823 |
GaN3O9 | |
MFCD00149723 | |
gallium iii nitrate hydrate, nitric acid, gallium salt, hydrate, ga.3no3.h2o, gallium nitrate nanoparticles, gallium-iii-nitrate hydrate, gallium iii nitrate hydrate, puratronic, gallium iii nitrate hydrate, puratronic metals basis, gallium iii nitrate hydrate, crystalline trace metals basis, gallium iii nitrate hydrate, crystals and lumps trace metals basis | |
[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H272-H315-H319
DOTInformation : Hazard Class: 5.1; Packaging Group: II
EINECSNumber : 236-815-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only