missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gallium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1115036

 View more versions of this product

Catalog No. AA1115036



Specifications

Specifications

Gallium(III) nitrate hydrate
(metals basis)
MFCD00149723
14,4346
Soluble in water.
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Ga+3]
273.75
255.73 (Anhydrous)
99.999%
Hygroscopic
110°C (decomposition)
69365-72-6
GaH2N3O10
UN1477
ga.3no3.h2o, gallium iii nitrate hydrate, gallium iii nitrate hydrate, crystalline trace metals basis, gallium iii nitrate hydrate, crystals and lumps trace metals basis, gallium iii nitrate hydrate, puratronic, gallium iii nitrate hydrate, puratronic metals basis, gallium nitrate nanoparticles, gallium-iii-nitrate hydrate, nitric acid, gallium salt, hydrate
YVFORYDECCQDAW-UHFFFAOYSA-N
gallium;trinitrate;hydrate
11550823
Crystalline Aggregates
Puratronic™
White
500g

Chemical Identifiers

69365-72-6
273.75
YVFORYDECCQDAW-UHFFFAOYSA-N
11550823
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Ga+3]
GaH2N3O10
MFCD00149723
ga.3no3.h2o, gallium iii nitrate hydrate, gallium iii nitrate hydrate, crystalline trace metals basis, gallium iii nitrate hydrate, crystals and lumps trace metals basis, gallium iii nitrate hydrate, puratronic, gallium iii nitrate hydrate, puratronic metals basis, gallium nitrate nanoparticles, gallium-iii-nitrate hydrate, nitric acid, gallium salt, hydrate
gallium;trinitrate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.

P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c

H272-H315-H319

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 236-815-5

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents