Learn More
Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), Thermo Scientific Chemicals
Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), ACROS Organics, Quantity: 100g, Packaging: Glass bottle, CAS: 69365-72-6, White, Grade: Trace Metal Basis, Melting Point: 110.0 deg.C, Molecular Formula: GaN3O9.xH2O, Molecular Weight: 255.74, Percent Purity: 99.9998%
$44.83 - $1520.30
Chemical Identifiers
CAS | 69365-72-6 |
---|---|
Molecular Formula | GaN3O9 |
Molecular Weight (g/mol) | 255.74 |
MDL Number | MFCD00149723 |
InChI Key | CHPZKNULDCNCBW-UHFFFAOYSA-N |
Synonym | gallium iii nitrate hydrate, nitric acid, gallium salt, hydrate, ga.3no3.h2o, gallium nitrate nanoparticles, gallium-iii-nitrate hydrate, gallium iii nitrate hydrate, puratronic, gallium iii nitrate hydrate, puratronic metals basis, gallium iii nitrate hydrate, crystalline trace metals basis, gallium iii nitrate hydrate, crystals and lumps trace metals basis |
PubChem CID | 11550823 |
SMILES | [Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC212440010
|
Thermo Scientific Chemicals
212440010 |
1 g | Glass bottle |
Each for $44.83
|
|
||||
AC212440050
|
Thermo Scientific Chemicals
212440050 |
5 g | Glass bottle |
Each for $95.67
|
|
||||
AC212440250
|
Thermo Scientific Chemicals
212440250 |
25 g | Glass bottle |
Each for $382.79
|
|
||||
AC212441000
|
Thermo Scientific Chemicals
212441000 |
100 g | Glass bottle |
Each for $1,520.30
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
69365-72-6 | |
255.74 | |
CHPZKNULDCNCBW-UHFFFAOYSA-N | |
11550823 |
GaN3O9 | |
MFCD00149723 | |
gallium iii nitrate hydrate, nitric acid, gallium salt, hydrate, ga.3no3.h2o, gallium nitrate nanoparticles, gallium-iii-nitrate hydrate, gallium iii nitrate hydrate, puratronic, gallium iii nitrate hydrate, puratronic metals basis, gallium iii nitrate hydrate, crystalline trace metals basis, gallium iii nitrate hydrate, crystals and lumps trace metals basis | |
[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
110.0°C | |
White | |
1 g | |
GaN3O9 | |
MFCD00149723 | |
gallium iii nitrate hydrate, nitric acid, gallium salt, hydrate, ga.3no3.h2o, gallium nitrate nanoparticles, gallium-iii-nitrate hydrate, gallium iii nitrate hydrate, puratronic, gallium iii nitrate hydrate, puratronic metals basis, gallium iii nitrate hydrate, crystalline trace metals basis, gallium iii nitrate hydrate, crystals and lumps trace metals basis | |
CHPZKNULDCNCBW-UHFFFAOYSA-N | |
gallium(3+) trinitrate | |
11550823 | |
99.9998% | |
Glass bottle |
69365-72-6 | |
Crystalline Powder, Crystals and/or Chunks | |
99.9998% | |
Ga(NO3)3·xH2O | |
14, 4350 | |
Solubility in water: soluble | |
[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
255.74 | |
255.74 | |
Trace Metal Basis | |
Gallium(III) nitrate hydrate |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only