missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gallium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04714803

 View more versions of this product

Catalog No. AA4714803



Description

Description

It is useful for the preparation of compounds of gallium(I) and gallium(II) and is reported as useful in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Specifications

345°C (sublimation)
212°C
13450-91-4
Beads
(metals basis)
MFCD00016107
gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basis
DWRNSCDYNYYYHT-UHFFFAOYSA-K
triiodogallane
83478
99.999%
Light and moisture sensitive
Gallium(III) iodide
Ultra dry
Yellow to Green
1 g
GaI3
UN3260
Decomposes in water.
[Ga](I)(I)I
450.436
450.43
Ampule
4.15 g/mL

Chemical Identifiers

13450-91-4
450.436
DWRNSCDYNYYYHT-UHFFFAOYSA-K
83478
[Ga](I)(I)I
GaI3
MFCD00016107
gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basis
triiodogallane
Safety and Handling

Safety and Handling

GHS H Statement
H334-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

P260-P264b-P271-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c

H314-H317-H334-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 236-611-6

TSCA : Yes

storageNote1 :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only