Learn More
Gallium(III) fluoride, anhydrous, 99.85% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03211206
Description
Gallium(III) fluoride reacts with mineral acids to give hydrofluoric acid. It is used in the preparation and study of optical fluorogallate glasses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
∼1000°C | |
800°C (sublimation) | |
7783-51-9 | |
Powder | |
(metals basis) | |
UN3260 | |
Insoluble in water. | |
99.85% | |
Moisture sensitive |
Gallium(III) fluoride | |
Anhydrous | |
White | |
5 g | |
MFCD00011021 | |
14,4354 | |
126.72 | |
Odorless | |
4.47 g/mL |
Chemical Identifiers
7783-51-9 | |
MFCD00011021 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 232-004-5
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only