Please login to your online account to display your discounted pricing

Gallium(III) chloride, ultra dry, 99.999% (metals basis), Alfa Aesar™

$75.95 - $75.95

Chemical Identifiers

Chemical Name or Material Gallium(III) chloride
CAS 13450-90-3
Physical Form Ultra Dry. Various Forms (Powder/Flake/Crystalline/Beads, Etc.)
Assay Percent Range 99.999% (Metal basis)
Molecular Formula GaCl3
Formula Weight 176.03
MDL Number MFCD00011018
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3569806

  SDS
alfa aesar
3569806*
5g Each for $75.95

AA3569814

  SDS
alfa aesar
3569814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3569822

  SDS
alfa aesar
3569822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.03
Physical Form Ultra Dry. Various Forms (Powder/Flake/Crystalline/Beads, Etc.)
Quantity 5g
Color White
DOT Information DOT Class: 8, Packing Group: II
Density 2.47g/cm3
EINECS Number 236-610-0
MDL Number MFCD00011018
Health Hazard 1 H301-H311-H332-H314-H318: Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.
Appearance Ultra dry. White powder/flake/crystalline/beads, etc.
Molecular Formula GaCl3
CAS 13450-90-3
Chemical Name or Material Gallium(III) chloride
Assay Percent Range 99.999% (Metal basis)
Safety and Handling

H301-H311-H332-H314-H318: Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.