missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gallium(III) chloride, ultra dry, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4387914

 View more versions of this product

Catalog No. AA4387914



Description

Description

  • Ampouled under argon
Specifications

Specifications

Gallium (III) chloride
13450-90-3
Cl3Ga
UN3260
gallium trichloride, gallium iii chloride, trichlorogallium, gallium chloride, gallium chloride gacl3, gallium 3+ chloride, gacl3, ccris 4492, gallium iii chloride, ultra dry, trichlorogalliue
Cl[Ga](Cl)Cl
176.073
176.03
99.999%
Ampule
201°C
78°C
Ultra dry
(metals basis)
MFCD00011018
14,4348
UPWPDUACHOATKO-UHFFFAOYSA-K
trichlorogallane
26010
Crystalline
2.47g/mL
Moisture Sensitive
White
25g

Chemical Identifiers

13450-90-3
176.073
UPWPDUACHOATKO-UHFFFAOYSA-K
26010
Cl[Ga](Cl)Cl
Cl3Ga
MFCD00011018
gallium trichloride, gallium iii chloride, trichlorogallium, gallium chloride, gallium chloride gacl3, gallium 3+ chloride, gacl3, ccris 4492, gallium iii chloride, ultra dry, trichlorogalliue
trichlorogallane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : LW9100000

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Documents