Please login to your online account to display your discounted pricing

Alfa Aesar™ Gallium(III) bromide, ultra dry, 99.998% (metals basis)

Chemical Identifiers

Chemical Name or Material Gallium(III) bromide
CAS 13450-88-9
Name Note Ultra dry
Formula Weight 309.45
MDL Number MFCD00016100
UN Number UN3260
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3569703

  SDS
alfa aesar™
3569703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3569706

  SDS
alfa aesar™
3569706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3569714

  SDS
alfa aesar™
3569714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 309.45
Packaging Ampule
Sensitivity Moisture Sensitive
Color White
CAS 13450-88-9
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Gallium(III) bromide
Physical Form Crystalline
Assay Percent Notes (metals basis)
MDL Number MFCD00016100
Density 3.69g/mL
Storage Note 1
  • Ampouled under argon
Name Note Ultra dry
TSCA TSCA
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Quantity 1g
Odor Odorless
UN Number UN3260
Melting Point 122°C
Boiling Point 279°C
Assay 99.998%

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

  • Ampouled under argon