Please login to your online account to display your discounted pricing

Alfa Aesar™ Gallium(II) chloride, ultra dry, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Gallium(II) chloride
CAS 24597-12-4
Name Note Ultra dry
Formula Weight 281.26
MDL Number MFCD00151526
UN Number UN3260
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3569603

  SDS
alfa aesar™
3569603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 281.26
Packaging Ampule
Sensitivity Air & Moisture Sensitive
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Odor Acidic
UN Number UN3260
Boiling Point 535°C
Storage Note 1
  • Ampouled under argon
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
CAS 24597-12-4
Name Note Ultra dry
Melting Point 164°C
Assay Percent Notes (metals basis)
Assay 99.999%
Quantity 1g
Chemical Name or Material Gallium(II) chloride
MDL Number MFCD00151526
Physical Form Powder
Color White
Health Hazard 1 Danger
TSCA TSCA

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

  • Ampouled under argon