missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gallium antimonide, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1293303

 View more versions of this product

Catalog No. AA1293303



Specifications

Specifications

Gallium antimonide
(metals basis)
UN1549
191.47
99.99%
6mm & down
1g
12064-03-8
MFCD00016101
gallium antimonide, stibinogallane, galliumantimonid, gallium antimonide, polycryst, gallium antimonide, single crystal substrate , <100>, diam. x thickness 2 in. x 0.5 mm
Pieces
Odorless
710°C

Chemical Identifiers

12064-03-8
MFCD00016101
gallium antimonide, stibinogallane, galliumantimonid, gallium antimonide, polycryst, gallium antimonide, single crystal substrate , <100>, diam. x thickness 2 in. x 0.5 mm
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.

TSCA : TSCA

Documents