missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gallium, 99.9999%, (trace metal basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 193070050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisSpecifications
Gray | |
29.7°C | |
2400.0°C | |
99.9999% | |
Ga | |
5 g | |
Solubility in water: insoluble. | |
[Ga] | |
69.72 | |
CHEBI:49631 | |
99.9999% | |
Plastic bottle |
Gallium | |
7440-55-3 | |
5.9000g/mL | |
Ga | |
MFCD00134045 | |
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom | |
GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
gallium | |
5360835 | |
69.72 | |
Trace Metal Basis | |
Solid and/or Liquid |
Chemical Identifiers
7440-55-3 | |
69.72 | |
GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
5360835 | |
gallium |
Ga | |
MFCD00134045 | |
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom | |
CHEBI:49631 | |
[Ga] |
Safety and Handling
Warning
Recommended Storage : 4°C
RUO – Research Use Only