missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gallic acid monohydrate, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC220760025

 View more versions of this product

Catalog No. AC220760025

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Gallic acid monohydrate
5995-86-8
≥99% (on dry substance)
(HO)3C6H2CO2H·H2O
MFCD00149098
15, 4375
Authentic
Plastic bottle
Off-white to beige
c.p.
Powder
C7H6O5·H2O
188.12
10, 470
3, 4, 5-Trihydroxybenzoic acid monohydrate
10% max. (120°C)
0.1% max.
2.5kg

Chemical Identifiers

5995-86-8
(HO)3C6H2CO2H·H2O
MFCD00149098
Gallic acid monohydrate, c.p.
C7H6O5·H2O
188.12
3, 4, 5-Trihydroxybenzoic acid monohydrate
≥99% (on dry substance)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning