missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gallic Acid 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 410860050

 View more versions of this product

Catalog No. AC410860050


 

Specifications Description & Specifications

Specifications

Gallic acid
149-91-7
≥97.5%
C7H6O5
MFCD00002510
1.6940g/mL
Glass Bottle
5g
98%
Powder
98%
170.12
3, 4, 5-Trihydroxybenzoic acid
Authentic
Light yellow to beige

Chemical Identifiers

149-91-7
C7H6O5
MFCD00002510
Gallic acid, 98%
98%
170.12
3, 4, 5-Trihydroxybenzoic acid
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 205-749-9

RTECSNumber : LW7525000

TSCA : TSCA