Please login to your online account to display your discounted pricing

Galileo Thermometer


Handcrafted and individually calibrated.

Manufacturer: fisher science education  B620000600

 View more versions of this product

Catalog No. S95614

  • / Each
  • / Case of 10 Each
 

Description & Specifications

Specifications

Accuracy 1°C
Length (English) Overall 13 in.
Height (Metric) 330mm
Temperature Range (Metric) 18° to 27°C
Height (English) 13 in.
Material Glass
Temperature Range (English) 64° to 80°F
Subdivision 4°C
Thermometer Fill Non Hazardous Liquid
Length (Metric) Overall 330mm