missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

H-B Instrument™ Galileo Thermometer

Handcrafted and individually calibrated.

Manufacturer:  H-B Instrument™ B620000400EMD

 View more versions of this product

Catalog No. S95613




Description

Description

  • Multi-colored spheres become more or less buoyant as the temperature of the liquid they are in changes
  • Each sphere differs in weight by only 0.002g
  • Filled with nonhazardous liquid
  • Standard models use optical crystal and are accurate to ±1°F
  • Economy models use laboratory-grade glass and are accurate to ±2°F

Specifications

Specifications

4°C
18° to 27°C
27°C
18°C
430mm
432mm
Non Hazardous Liquid
64° to 80°F
80°F
64°F
17 in.
17 in.
Glass
1°C
Documents