Learn More
Galena, naturally occurring mineral, grains, approximately 0.06-0.19in, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04264014
Description
Galena is used in infrared detectors, photo optic applications, slip property modifier used in friction industry for enhancing heat conduction and regulating friction coefficient, used as a semiconductor. Used in electronic devices and Infrared sensor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Galena | |
Black | |
Powder, Flake, Crystalline, Beads, Lump, Etc. | |
PbS | |
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide | |
FILXVLQXZYXBSJ-UHFFFAOYSA-N | |
sulfanylidenelead | |
14819 | |
Odorless |
naturally occurring mineral, grains, approximately 0.06-0.19 in. | |
1314-87-0 | |
25 g | |
MFCD00016280 | |
Soluble in water (0.00086g/L) and acid. Insoluble in alcohol,and potassium hydroxide. | |
[S--].[Pb++] | |
239.30 | |
239.25 |
Chemical Identifiers
1314-87-0 | |
239.30 | |
FILXVLQXZYXBSJ-UHFFFAOYSA-N | |
14819 | |
[S--].[Pb++] |
PbS | |
MFCD00016280 | |
lead ii sulfide, lead sulfide, galena, lead monosulfide, plumbous sulfide, lead 2+ sulfide, natural galena, lead sulphide, natural lead sulfide | |
sulfanylidenelead |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H373
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 215-246-6
RTECSNumber : OG4550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only