Please login to your online account to display your discounted pricing

Galectin-3 Rat anti-Mouse, eFluor 660, Clone: eBioM3/38 (M3/38), eBioscience™

Rat Monoclonal Antibody

$166.75 - $359.60

Specifications

Antigen Galectin-3
Clone eBioM3/38 (M3/38)
Host Species Rat
Gene Alias MAC2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124381

 
life technologies
LS50530180
25μg Each for $166.75

501124382

 
life technologies
LS50530182
100μg Each for $359.60
Description & Specifications

Specifications

Antigen Galectin-3
Clone eBioM3/38 (M3/38)
Host Species Rat
Gene Alias MAC2
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eBioM3/38 (M3/38) antibody reacts with mouse Galectin-3 (Mac-2). Galectins are a family of animal lectins which appear to exhibit a variety of biological functions. Galectin 3 (aka LGALS3, galactose-specific soluble lectin 3, Mac-2, L-29) is one of the more extensively studied members of this family and is a 30kDa beta-galactoside-binding protein. Due to a C-terminal carbohydrate binding site, Galectin 3 is capable of binding IgE and mammalian cell surfaces only when homodimerized or homo-oligomerized. Galectin 3 is normally distributed in epithelia of many organs and various inflammatory cells, including macrophages, as well as dendritic cells and Kupffer cells. The expression of this lectin is up-regulated during inflammation, cell proliferation, cell differentiation and through trans-activation by viral proteins.

This monoclonal antibody eBioM3/38 crossreacts to human Galectin-3 (LGALS3).