missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Galectin-3 Rat anti-Human, Mouse, PE, Clone: eBioM3/38 (M3/38), eBioscience™

Rat Monoclonal Antibody

$201.55 - $433.55

Specifications

Antigen Galectin-3
Clone eBioM3/38 (M3/38)
Host Species Rat
Gene Alias MAC2
Species Reactivity Human, Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010815 Life Technologies
LS12530181
50μg Each for $201.55
5010816 Life Technologies
LS12530182
100μg Each for $288.55
5010817 Life Technologies
LS12530183
200μg Each for $433.55
Specifications

Description

The eBioM3/38 antibody reacts with mouse Galectin-3 (Mac-2). Galectins are a family of animal lectins which appear to exhibit a variety of biological functions. Galectin 3 (aka LGALS3, galactose-specific soluble lectin 3, Mac-2, L-29) is one of the more extensively studied members of this family and is a 30kDa beta-galactoside-binding protein. Due to a C-terminal carbohydrate binding site, Galectin 3 is capable of binding IgE and mammalian cell surfaces only when homodimerized or homo-oligomerized. Galectin 3 is normally distributed in epithelia of many organs and various inflammatory cells, including macrophages, as well as dendritic cells and Kupffer cells. The expression of this lectin is up-regulated during inflammation, cell proliferation, cell differentiation and through trans-activation by viral proteins.

This monoclonal antibody eBioM3/38 crossreacts to human Galectin-3 (LGALS3).
Description & Specifications

Specifications

Galectin-3
Rat
Human, Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioM3/38 (M3/38)
MAC2
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL