missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
GALE, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002582B01P
Description
Sequence: MAEKVLVTGGAGYIGSHTVLELLEAGYLPVVIDNFHNAFRGGGSLPESLRRVQELTGRSVEFEEMDILDQGALQRLFKKYSFMAVIHFAGLKAVGESVQKPLDYYRVNLTGTIQLLEIMKAHGVKNLVFSSSATVYGNPQYLPLDEAHPTGGCTNPYGKSKFFIEEMIRDLCQADKTWNAVLLRYFNPTGAHASGCIGEDPQGIPNNLMPYVSQVAIGRREALNVFGNDYDTEDGTGVRDYIHVVDLAKGHIAALRKLKEQCGCRIYNLGTGTGYSVLQMVQAMEKASGKKIPYKVVARREGDVAACYANPSLAQEELGWTAALGLDRMCEDLWRWQKQNPSGFGTQASpecifications
GALE | |
Polyclonal | |
In 1x PBS, pH 7.4 | |
NM_000403.3 | |
GALE | |
GALE (NP_000394.2, 1 a.a. ∼ 348 a.a) full-length human protein. | |
RUO | |
2582 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
UDP-galactose-4-epimerase | |
FLJ95174/FLJ97302/SDR1E1 | |
Mouse | |
50 μg | |
Primary | |
Human | |
Antibody |