Please login to your online account to display your discounted pricing

Alfa Aesar™ Galantide

For cellular and molecular biology applications

Specifications

Product Type Galantide
Molecular Weight 2199.60
Storage Requirements -30° to -10°C
Sequence Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66575EXD

 
alfa aesar™
J66575EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66575MCR

 
alfa aesar™
J66575MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Galantide
Molecular Weight 2199.60
Storage Requirements -30° to -10°C
Sequence Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC