missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Galanthamine hydrobromide

Centrally active and long-acting acetylcholinesterase inhibitor

$127.85 - $1,023.97

Chemical Identifiers

CAS 1953-04-4
Molecular Formula C17H22BrNO3
Molecular Weight (g/mol) 368.27
MDL Number MFCD00067672
InChI Key QORVDGQLPPAFRS-XPSHAMGMSA-N
Synonym galanthamine hydrobromide, galantamine hydrobromide, galanthamine hydrobromide, nivalin, razadyne, mj4ptd2vvw, unii-mj4ptd2vvw, nivaline pharmaceutical, galanthamine hbr, lycoremine hydrobromide
PubChem CID 121587
SMILES CN1CCC23C=CC(CC2OC4=C(C=CC(=C34)C1)OC)O.Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62482MF View Documents Alfa Aesar™
J62482MF
50mg Each for $127.85
 
AAJ6248203 View Documents Alfa Aesar™
J6248203
1g Each for $1,023.97
 
Specifications

Chemical Identifiers

1953-04-4
368.27
QORVDGQLPPAFRS-XPSHAMGMSA-N
121587
C17H22BrNO3
MFCD00067672
galanthamine hydrobromide, galantamine hydrobromide, galanthamine hydrobromide, nivalin, razadyne, mj4ptd2vvw, unii-mj4ptd2vvw, nivaline pharmaceutical, galanthamine hbr, lycoremine hydrobromide
CN1CCC23C=CC(CC2OC4=C(C=CC(=C34)C1)OC)O.Br

Specifications

White
1953-04-4
MFCD00067672
14,4340
Soluble in water or DMSO
CN1CCC23C=CC(CC2OC4=C(C=CC(=C34)C1)OC)O.Br
121587
Powder
50mg
C17H22BrNO3
UN2811
galanthamine hydrobromide, galantamine hydrobromide, galanthamine hydrobromide, nivalin, razadyne, mj4ptd2vvw, unii-mj4ptd2vvw, nivaline pharmaceutical, galanthamine hbr, lycoremine hydrobromide
QORVDGQLPPAFRS-XPSHAMGMSA-N
368.27
368.27
Galanthamine hydrobromide
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : DF8075000

Recommended Storage : −30° to −10°C