Please login to your online account to display your discounted pricing

Alfa Aesar™ Galanin (2-11)


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66796EXD

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66796EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Galanin (2-11)
Molecular Weight 1136.30
Sequence Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC
Quantity 0.5mg
Storage Requirements -30° to -10°C