Please login to your online account to display your discounted pricing

Alfa Aesar™ Galanin (1-16), Porcine, rat

For cellular and molecular biology applications

Specifications

Product Type Galanin (1-16), porcine, rat
Molecular Weight 1669.92
Storage Requirements -30° to -10°C
Sequence Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-His-Ala-Ile
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66716LB0

 
alfa aesar™
J66716LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66716MA

 
alfa aesar™
J66716MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Galanin (1-16), porcine, rat
Molecular Weight 1669.92
Storage Requirements -30° to -10°C
Sequence Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-His-Ala-Ile
Format Solid
Regulatory Status RUO
Purification Method HPLC