Please login to your online account to display your discounted pricing

Alfa Aesar™ Galanin (1-13)-Spantide I

For cellular and molecular biology applications

Specifications

Product Type Galanin (1-13)-Spantide I
Molecular Weight 2828.34
Storage Requirements -30° to -10°C
Sequence Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-D-Arg-Pro-Lys-Pro-Gln-Gln-D-Trp-Phe-D-Trp-Leu-Leu-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66022LB0

 
alfa aesar™
J66022LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66022MCR

 
alfa aesar™
J66022MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Galanin (1-13)-Spantide I
Molecular Weight 2828.34
Storage Requirements -30° to -10°C
Sequence Gly-Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-D-Arg-Pro-Lys-Pro-Gln-Gln-D-Trp-Phe-D-Trp-Leu-Leu-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC