Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ β -Galactosidase Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting, immunoprecipitation, and protein purification applications.

$276.82 - $1,519.06

Specifications

Antigen beta-Galactosidase Ab-1
Clone BGAL01
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity    

MS1329B

 
thermo scientific™ lab vision™
MS1329B
200μg/mL Biotin 1mL Each for $1,519.06

MS1329B0

 
thermo scientific™ lab vision™
MS1329B0
200μg/mL Biotin 100µL Each for $374.45

MS1329B1

 
thermo scientific™ lab vision™
MS1329B1
200μg/mL Biotin 500µL Each for $1,007.10

MS1329P

 
thermo scientific™ lab vision™
MS1329P
200μg/mL Unlabeled 1mL Each for $1,179.94

MS1329P0

 
thermo scientific™ lab vision™
MS1329P0
200μg/mL Unlabeled 100µL Each for $276.82

MS1329P1

 
thermo scientific™ lab vision™
MS1329P1
200μg/mL Unlabeled 500µL Each for $774.59

MS1329P1ABX

 
thermo scientific™ lab vision™
MS1329P1ABX
1mg/mL Unlabeled 100µL Each for $860.16

MS1329PABX

 
thermo scientific™ lab vision™
MS1329PABX
1mg/mL Unlabeled 200µL Each for $1,278.80
Description & Specifications

Specifications

Antigen beta-Galactosidase Ab-1
Clone BGAL01
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Cross Reactivity All
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Epitope Tags
Type Primary
Immunogen E. coli beta-galactosidase
Isotype IgG1

Ensure accurate, reproducible results in western blotting, immunoprecipitation, and protein purification applications with Thermo Scientific™ β-Galactosidase Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: BGAL01

Isotype: IgG1

Species Reactivity: Proteins from all species which are tagged with β-galactosidase.

Epitope: Not determined

Immunogen: E. coli β-galactosidase

Molecular Weight: 116kDa

Positive Control: Purified recombinant β-galactosidase protein or transfected cell lines expressing β-galactosidase-tagged proteins

Cellular Localization: Depends upon the localization of the parent protein tagged with β-galactosidase

Recommended for:

  • Western Blotting
  • Immunoprecipitation
  • Protein purification

Note:

USA: RUO; Int'l: RUO