Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ β -Galactosidase Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in western blotting, immunoprecipitation, and protein purification applications.

Manufacturer: thermo scientific™ lab vision™  MS1329PABX

 View more versions of this product

Catalog No. MS1329PABX

  • / Each
 

Description & Specifications

Specifications

Quantity 200µL
Antigen beta-Galactosidase Ab-1
Format Concentrated
Regulatory Status RUO
Host Species Mouse
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1
Cross Reactivity All
Clone BGAL01
Immunogen E. coli beta-galactosidase
Classification Monoclonal
Conjugate Unlabeled
Concentration 1mg/mL
Type Primary
Research Discipline Epitope Tags

Ensure accurate, reproducible results in western blotting, immunoprecipitation, and protein purification applications with Thermo Scientific™ β-Galactosidase Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: BGAL01

Isotype: IgG1

Species Reactivity: Proteins from all species which are tagged with β-galactosidase.

Epitope: Not determined

Immunogen: E. coli β-galactosidase

Molecular Weight: 116kDa

Positive Control: Purified recombinant β-galactosidase protein or transfected cell lines expressing β-galactosidase-tagged proteins

Cellular Localization: Depends upon the localization of the parent protein tagged with β-galactosidase

Recommended for:

  • Western Blotting
  • Immunoprecipitation
  • Protein purification

Note:

USA: RUO; Int'l: RUO