missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gadolinium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$261.12 - $449.24
Chemical Identifiers
CAS | 7440-54-2 |
---|---|
Molecular Formula | Gd |
Molecular Weight (g/mol) | 157.25 |
MDL Number | MFCD00011022 |
InChI Key | UIWYJDYFSGRHKR-UHFFFAOYSA-N |
PubChem CID | 23982 |
ChEBI | CHEBI:33375 |
IUPAC Name | gadolinium |
SMILES | [Gd] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
PLGD2-2Y
|
SPEX CertiPrep Group LLC
PLGD22Y |
125 mL | Triple-leached LDPE bottle |
Each for $261.12
|
|
||||
PLGD2-2X
|
SPEX CertiPrep Group LLC
PLGD22X |
500 mL | Triple-leached LDPE bottle |
Each for $449.24
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Gd | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-54-2 | |
157.25 | |
UIWYJDYFSGRHKR-UHFFFAOYSA-N | |
CHEBI:33375 | |
[Gd] |
Gd | |
MFCD00011022 | |
23982 | |
gadolinium |
Specifications
7440-54-2,7697-37-2,7732-18-5 | |
125 mL | |
0.1,2.0,97.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Gd | |
UIWYJDYFSGRHKR-UHFFFAOYSA-N | |
gadolinium | |
23982 | |
Assurance | |
2% HNO3 |
Triple-leached LDPE bottle | |
0.1,2.0,97.9 | |
Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00011022 | |
[Gd] | |
157.25 | |
CHEBI:33375 | |
Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mL PLGD2-2Y, -2X 10,000 PLGD2-3Y, -3X |
|
Gadolinium Standard |
Safety and Handling
Causes severe skin burns and eye damage
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only