Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium sputtering target, 76.2mm dia x 3.18mm thick

$1,583.61 - $1,583.61

Chemical Identifiers

Chemical Name or Material Gadolinium sputtering target
CAS 7440-54-2
Name Note 76.2mm dia. x 3.18mm thick
MDL Number MFCD00011022
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45929KS

 
alfa aesar™
45929KS
1 ea. Each for $1,583.61
Description & Specifications

Specifications

Quantity 1 ea.
Chemical Name or Material Gadolinium sputtering target
TSCA TSCA
Name Note 76.2mm dia. x 3.18mm thick
MDL Number MFCD00011022
CAS 7440-54-2